Learn More
Biomatik Corporation Human Free Fatty Acid Receptor 1 (FFAR1) ELISA Kit
Supplier:  Biomatik Corporation EKU04271
Encompass_Preferred
Human Free Fatty Acid Receptor 1 (FFAR1) ELISA Kit for detecting FFAR1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O14842. Gene ID: 2864. Target Synonyms: GPR40; FFA1R; G Protein-Coupled Receptor 40. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2400