Learn More
Biomatik Corporation Human Fucosidase Alpha L1, Tissue (FUCa1) ELISA Kit
Supplier:  Biomatik Corporation EKU04280
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.123ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P04066. Target Synonyms: A-FU; aFU; Alpha-L-Fucosidase; Alpha-L-fucoside fucohydrolase 1. Restriction: For Research Use Only.
Catalog No. 50-229-2403