Learn More
Biomatik Corporation Rat Galectin 9 (GAL9) ELISA Kit
Supplier:  Biomatik Corporation EKU04329
Encompass_Preferred
Rat Galectin 9 (GAL9) ELISA Kit for detecting GAL9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97840. Gene ID: 25476. Target Synonyms: LGALS9; Ecalectin; Lectin,Galactoside-Binding Soluble 9; Tumor antigen HOM-HD-21. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2417