Learn More
Biomatik Corporation Human Gap Junction Protein Beta 1 (GJb1) ELISA Kit
Catalog No. 502292419
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.108ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P08034. Target Synonyms: CMTX1; CX32; GJ-B1; Connexin 32; Charcot-Marie-Tooth Neuropathy,X-Linked; GAP junction 28 kDa liver protein. Restriction: For Research Use Only.
Catalog No. 50-229-2419 Supplier Biomatik Corporation Supplier No. EKU04339
Add to Cart
Add to Cart
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.108ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P08034. Target Synonyms: CMTX1; CX32; GJ-B1; Connexin 32; Charcot-Marie-Tooth Neuropathy,X-Linked; GAP junction 28 kDa liver protein. Restriction: For Research Use Only.