Learn More
Biomatik Corporation Human Glucocorticoid Receptor Beta (GRb) ELISA Kit
Supplier:  Biomatik Corporation EKU04398
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.112ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P04150 & P04150-2. Target Synonyms: GR-B. Restriction: For Research Use Only.
Catalog No. 50-229-2427