Learn More
Biomatik Corporation Rat Glutamate Cysteine Ligase, Modifier Subunit (GCLM) ELISA Kit
Supplier:  Biomatik Corporation EKU04436
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: GLCLR; GECS; GCL; Gamma Glutamylcysteine Synthetase; Glutamate Cysteine Ligase Regulatory; Gamma-ECS regulatory subunit; Glutamate cysteine ligase modifier subunit. Restriction: For Research Use Only.
Catalog No. 50-229-2434