Learn More
Biomatik Corporation Pig Glutathione S Transferase Pi (GSTp) ELISA Kit
Supplier:  Biomatik Corporation EKU04498
Encompass_Preferred
Pig Glutathione S Transferase Pi (GSTp) ELISA Kit for detecting GSTp in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.56-100ng/ml. Sensitivity: 0.63ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P80031. Target Synonyms: GSTP1; GST-P1; DFN7; FAEES3; GST3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2447