Learn More
Biomatik Corporation Human Glycoprotein IIb/IIIa (GPIIb/IIIa) ELISA Kit
Supplier:  Biomatik Corporation EKU04532
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.059ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: GP-IIb/IIIa. Restriction: For Research Use Only.
Catalog No. 50-229-2454