Learn More
Biomatik Corporation Mouse Growth Arrest Specific Protein 6 (GAS6) ELISA Kit
Supplier:  Biomatik Corporation EKU04578
Encompass_Preferred
Mouse Growth Arrest Specific Protein 6 (GAS6) ELISA Kit for detecting GAS6 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61592. Gene ID: 14456. Target Synonyms: AXLLG; AXSF; AXL Stimulatory Factor; AXL receptor tyrosine kinase ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2464