Learn More
Biomatik Corporation Mouse Bone Morphogenetic Protein 9 (BMP9) ELISA Kit
Supplier:  Biomatik Corporation EKU04592
Encompass_Preferred
Mouse Bone Morphogenetic Protein 9 (BMP9) ELISA Kit for detecting BMP9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.12-5000pg/ml. Sensitivity: 28pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WV56. Gene ID: 12165. Target Synonyms: GDF2; Growth Differentiation Factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2466