Learn More
Biomatik Corporation Rat Growth Differentiation Factor 5 (GDF5) ELISA Kit
Supplier:  Biomatik Corporation EKU04599
Encompass_Preferred
Rat Growth Differentiation Factor 5 (GDF5) ELISA Kit for detecting GDF5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: M0RAY4. Gene ID: 252835. Target Synonyms: CDMP1; LAP4; SYNS2; BMP14; Radotermin; Cartilage-Derived Morphogenetic Protein-1; Bone morphogenetic protein 14; Lipopolysaccharide-associated protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2469