Learn More
Biomatik Corporation Mouse Growth Factor, Augmenter Of Liver Regeneration (GFER) ELISA Kit
Supplier:  Biomatik Corporation EKU04604
Encompass_Preferred
Mouse Growth Factor, Augmenter Of Liver Regeneration (GFER) ELISA Kit for detecting GFER in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P56213. Gene ID: 11692. Target Synonyms: ERV1; ALR; HERV1; HPO; HPO1; HPO2; HSS; FAD-linked sulfhydryl oxidase ALR; Augmenter of liver regeneration. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2471