Learn More
Biomatik Corporation Human alpha-amylase (AMY1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC32542
Encompass_Preferred
The Human alpha-amylase (AMY1) ELISA Kit allows for quantitative determination of alpha-amylase in serum, saliva, tissue homogenates. Species Reactivity: Human. Detection Range: 7.8 IU/mL-500 IU/mL. Sensitivity: 1.95 IU/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P04745. Restriction: For Research Use Only.
Catalog No. 50-149-7670