Learn More
Biomatik Corporation Dog Haptoglobin (Hpt) ELISA Kit
Supplier:  Biomatik Corporation EKU04633
Encompass_Preferred
Dog Haptoglobin (Hpt) ELISA Kit for detecting Hpt in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 3.12-200ng/ml. Sensitivity: 1.23ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19006. Target Synonyms: HP; Hp2-Alpha; Alpha-2-Macroglobulin; Zonulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2473