Learn More
Biomatik Corporation Rat Heat Shock Protein Beta 6 (HSPb6) ELISA Kit
Supplier:  Biomatik Corporation EKU04690
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.068ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P97541. Target Synonyms: HSP20; Heat Shock Protein 20 kD; Heat Shock Protein B6, Alpha Crystallin Related; Heat shock 20 kDa-like protein p20. Restriction: For Research Use Only.
Catalog No. 50-229-2480