Learn More
Biomatik Corporation Bovine Hepatocyte Growth Factor (HGF) ELISA Kit
Supplier:  Biomatik Corporation EKU04749
Encompass_Preferred
Bovine Hepatocyte Growth Factor (HGF) ELISA Kit for detecting HGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 15.62-1000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q76BS1. Gene ID: 282879. Target Synonyms: HGF; F-TCF; HGFB; HPTA; SF; Scatter Factor; Hepapoietin A; Fibroblast-Derived Tumor Cytotoxic Factor; Lung Fibroblast-Derived Mitogen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2488