Learn More
Biomatik Corporation Human Heterogeneous Nuclear Ribonucleoprotein A2/B1 (HNRPA2B1) ELISA Kit
Supplier:  Biomatik Corporation EKU04769
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P22626. Target Synonyms: hnRNP-A2/B1; hnRNPA2B1; RA33; HnRNPA2/B1. Restriction: For Research Use Only.
Catalog No. 50-229-2490