Learn More
Biomatik Corporation Human Histatin 3 (HTN3) ELISA Kit
Supplier:  Biomatik Corporation EKU04802
Encompass_Preferred
Human Histatin 3 (HTN3) ELISA Kit for detecting HTN3 in saliva, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.63ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15516. Gene ID: 3347. Target Synonyms: HIS2; HTN2; HTN5; Hst; PB; Histidine-rich protein 3; Basic histidine-rich protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2493