Learn More
Biomatik Corporation Mouse Hydroxyacyl Coenzyme A Dehydrogenase Beta (HADHb) ELISA Kit
Supplier:  Biomatik Corporation EKU04853
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 1.56-100ng/mL. Sensitivity: 0.61ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q99JY0. Target Synonyms: MTPB; Mitochondrial Trifunctional Protein,Beta Subunit; 3-Ketoacyl-Coenzyme A Thiolase/Enoyl-Coenzyme A Hydratase(Trifunctional Protein)Beta Subunit; Beta-ketothiolase. Restriction: For Research Use Only.
Catalog No. 50-229-2504