Learn More
Biomatik Corporation Rat Indoleamine-2,3-Dioxygenase (IDO) ELISA Kit
Supplier:  Biomatik Corporation EKU04918
Encompass_Preferred
Rat Indoleamine-2,3-Dioxygenase (IDO) ELISA Kit for detecting IDO in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ERD9. Gene ID: 66029. Target Synonyms: CD107B; INDO; Indoleamine-Pyrrole 2,3 Dioxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2511