Learn More
Biomatik Corporation Human Inhibin Beta B (INHbB) ELISA Kit
Supplier:  Biomatik Corporation EKU04934
Encompass_Preferred
Human Inhibin Beta B (INHbB) ELISA Kit for detecting INHbB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09529. Gene ID: 3625. Target Synonyms: INH-BB; Activin AB Beta Polypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2513