Learn More
Biomatik Corporation Mouse Insulin Degrading Enzyme (IDE) ELISA Kit
Supplier:  Biomatik Corporation EKU04961
Encompass_Preferred
Mouse Insulin Degrading Enzyme (IDE) ELISA Kit for detecting IDE in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JHR7. Target Synonyms: Insulysin; Insulin Protease; Abeta-degrading protease; Insulinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2517