Learn More
Biomatik Corporation Bovine Insulin Like Growth Factor 2 (IGF2) ELISA Kit
Supplier:  Biomatik Corporation EKU04975
Encompass_Preferred
Bovine Insulin Like Growth Factor 2 (IGF2) ELISA Kit for detecting IGF2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.21ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07456. Gene ID: 281240. Target Synonyms: INSIGF; IGF-II; Somatomedin A; Preptin; Insulin-like growth factor II Ala-25 Del. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2522