Learn More
Biomatik Corporation Human Integrin Alpha 1 (ITGa1) ELISA Kit
Supplier:  Biomatik Corporation EKU05026
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 78-5,000pg/mL. Sensitivity: 32pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P56199. Target Synonyms: CD49-A; CD49a; ITG-A1; VLA1; CD49 antigen-like family member A; Laminin and collagen receptor. Restriction: For Research Use Only.
Catalog No. 50-229-2527