Learn More
Biomatik Corporation Rabbit Intercellular Adhesion Molecule 1 (ICAM1) ELISA Kit
Supplier:  Biomatik Corporation EKU05064
Encompass_Preferred
Rabbit Intercellular Adhesion Molecule 1 (ICAM1) ELISA Kit for detecting ICAM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 6.25-400pg/ml. Sensitivity: 2.99pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: U3KPG6. Target Synonyms: CD54; BB2; P3.58; Human Rhinovirus Receptor; Major group rhinovirus receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2533