Learn More
Biomatik Corporation Human Interferon Induced Helicase C Domain Containing Protein 1 (IFIH1) ELISA Kit
Supplier:  Biomatik Corporation EKU05113
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.115ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9BYX4. Target Synonyms: Hlcd; IDDM19; MDA5; Helicard; Clinically amyopathic dermatomyositis autoantigen 140 kDa; Melanoma differentiation-associated protein 5; Murabutide down-regulated protein. Restriction: For Research Use Only.
Catalog No. 50-229-2538