Learn More
Biomatik Corporation Mouse Interferon Regulatory Factor 5 (IRF5) ELISA Kit
Supplier:  Biomatik Corporation EKU05124
Encompass_Preferred
Mouse Interferon Regulatory Factor 5 (IRF5) ELISA Kit for detecting IRF5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P56477. Gene ID: 27056. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2541