Learn More
Biomatik Corporation Bovine Interleukin 12A (IL12A) ELISA Kit
Supplier:  Biomatik Corporation EKU05196
Encompass_Preferred
Bovine Interleukin 12A (IL12A) ELISA Kit for detecting IL12A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P54349. Gene ID: 281856. Target Synonyms: p35; IL12-A; CLMF1; NFSK; NKSF1; Natural Killer Cell Stimulatory Factor 1; Cytotoxic Lymphocyte Maturation Factor 1; NF Cell Stimulatory Factor Chain 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2553