Learn More
Biomatik Corporation Rat Interleukin 19 (IL19) ELISA Kit
Supplier:  Biomatik Corporation EKU05265
Encompass_Preferred
Rat Interleukin 19 (IL19) ELISA Kit for detecting IL19 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZFI7. Target Synonyms: MDA1; ZMDA1, IL-10C, NG.1; Melanoma Differentiation Associated Protein-Like Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2563