Learn More
Biomatik Corporation Mouse IQ Motif Containing GTPase Activating Protein 1 (IQGAP1) ELISA Kit
Supplier:  Biomatik Corporation EKU05396
Encompass_Preferred
Mouse IQ Motif Containing GTPase Activating Protein 1 (IQGAP1) ELISA Kit for detecting IQGAP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JKF1. Gene ID: 29875. Target Synonyms: HUMORFA01; SAR1; p195. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2581