Learn More
Biomatik Corporation Rat Janus Kinase 3 (JAK3) ELISA Kit
Supplier:  Biomatik Corporation EKU05417
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Rat. Detection Range: 15.62-1,000pg/mL. Sensitivity: 6.2pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: JAKL; L-JAK; LJAK; A Protein Tyrosine Kinase,Leukocyte; Leukocyte janus kinase; Tyrosine-protein kinase JAK3. Restriction: For Research Use Only.
Catalog No. 50-229-2587