Learn More
Biomatik Corporation Human Kallikrein 11 (KLK11) ELISA Kit
Supplier:  Biomatik Corporation EKU05431
Encompass_Preferred
Human Kallikrein 11 (KLK11) ELISA Kit for detecting KLK11 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UBX7. Gene ID: 11012. Target Synonyms: TLSP; hK11; Hippostatin; Kallikrein-Related Peptidase 11; Serine protease 20; Trypsin-like protease. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2589