Learn More
Biomatik Corporation Human Leprecan Like Protein 1 (LEPREL1) ELISA Kit
Supplier:  Biomatik Corporation EKU05588
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.063ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: MLAT4; P3H2; Prolyl 3-Hydroxylase 2; Myxoid liposarcoma-associated protein 4. Restriction: For Research Use Only.
Catalog No. 50-229-2612