Learn More
Biomatik Corporation Human Lumican (LUM) ELISA Kit
Supplier:  Biomatik Corporation EKU05689
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.119ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P51884. Target Synonyms: LDC; SLRR2D; KSPG lumican; Keratan sulfate proteoglycan lumican. Restriction: For Research Use Only.
Catalog No. 50-229-2626