Learn More
Biomatik Corporation Rat Lymphotoxin Beta (LTb) ELISA Kit
Supplier:  Biomatik Corporation EKU05708
Encompass_Preferred
Rat Lymphotoxin Beta (LTb) ELISA Kit for detecting LTb in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6MG45. Gene ID: 361795. Target Synonyms: LT-B; p33; TNFC; TNF-C; TNFSF3; Tumor necrosis factor ligand superfamily member 3; Tumor Necrosis Factor C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2628