Learn More
Biomatik Corporation Human Lysophosphatidic Acid Receptor 1 (LPAR1) ELISA Kit
Supplier:  Biomatik Corporation EKU05711
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.115ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: EDG2; Gpcr26; LPA1; Mrec1.3; edg-2; rec.1.3; vzg-1; Endothelial Differentiation,Lysophosphatidic Acid G-Protein-Coupled Receptor 2. Restriction: For Research Use Only.
Catalog No. 50-229-2630