Learn More
Biomatik Corporation Human Lysophosphatidylcholine Acyltransferase 1 (LPCAT1) ELISA Kit
Supplier:  Biomatik Corporation EKU05714
Encompass_Preferred
Human Lysophosphatidylcholine Acyltransferase 1 (LPCAT1) ELISA Kit for detecting LPCAT1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.22ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8NF37. Gene ID: 79888. Target Synonyms: AYTL2; PFAAP3; Acyltransferase Like 2; 1-alkylglycerophosphocholine O-acetyltransferase; Phosphonoformate immuno-associated protein 3; Acetyl-CoA:lyso-PAF acetyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2632