Learn More
Biomatik Corporation Human Major Histocompatibility Complex Class II DQ Beta 1 (MHCDQb1) ELISA Kit
Supplier:  Biomatik Corporation EKU05774
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.125ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: HLA-DQB1; HLADQb1; CELIAC1; HLA-DQB; IDDM1; HLA Class II Histocompatibility Antigen,DQ Beta 1 Chain. Restriction: For Research Use Only.
Catalog No. 50-229-2638