Learn More
Biomatik Corporation Mouse Mannose Binding Lectin (MBL) ELISA Kit
Supplier:  Biomatik Corporation EKU05790
Encompass_Preferred
Mouse Mannose Binding Lectin (MBL) ELISA Kit for detecting MBL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.072ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P41317. Gene ID: 17195. Target Synonyms: MBL2; COLEC1; HSMBPC; MBP1; MBP; Collectin-1; Mannose-binding protein C; Mannan Binding Protein; Mannose-Binding Lectin (Protein C) 2,Soluble (Opsonic Defect). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2640