Learn More
Biomatik Corporation Rabbit Matrix Metalloproteinase 1 (MMP1) ELISA Kit
Supplier:  Biomatik Corporation EKU05822
Encompass_Preferred
Rabbit Matrix Metalloproteinase 1 (MMP1) ELISA Kit for detecting MMP1 in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.127ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13943. Gene ID: 100009110. Target Synonyms: CLGN; CLG1; Collagenase; Interstitial Collagenase; Vertebrate Collagenase; Fibroblast Collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2650