Learn More
Biomatik Corporation Human Matrix Metalloproteinase 16 (MMP16) ELISA Kit
Supplier:  Biomatik Corporation EKU05845
Encompass_Preferred
Human Matrix Metalloproteinase 16 (MMP16) ELISA Kit for detecting MMP16 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51512. Gene ID: 4325. Target Synonyms: MT3-MMP; MMP16; MMP-X2; MT-MMP2; MT-MMP3; Membrane-type matrix metalloproteinase 3; Membrane-type-3 matrix metalloproteinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2653