Learn More
Biomatik Corporation Human Matrix Metalloproteinase 25 (MMP25) ELISA Kit
Supplier:  Biomatik Corporation EKU05857
Encompass_Preferred
Human Matrix Metalloproteinase 25 (MMP25) ELISA Kit for detecting MMP25 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NPA2. Gene ID: 64386. Target Synonyms: MT6-MMP; MMP25; MMPL1; MT-MMP6; Matrix Metallopeptidase Like 1; Leukolysin; Membrane-type matrix metalloproteinase 6; Membrane-type-6 matrix metalloproteinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2657