Learn More
Biomatik Corporation Human Melanoma Inhibitory Activity Protein 1 (MIA1) ELISA Kit
Supplier:  Biomatik Corporation EKU05896
Encompass_Preferred
Human Melanoma Inhibitory Activity Protein 1 (MIA1) ELISA Kit for detecting MIA1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.81-500pg/ml. Sensitivity: 3.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q16674. Gene ID: 8190. Target Synonyms: CD-RAP; Melanoma-derived growth regulatory protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2666