Learn More
Biomatik Corporation Human Mitogen Activated Protein Kinase Kinase 1 (MAP2K1) ELISA Kit
Supplier:  Biomatik Corporation EKU05977
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q02750. Target Synonyms: MAPKK1; MEK1; MKK1; PRKMK1; Dual specificity mitogen-activated protein kinase kinase 1; ERK activator kinase 1. Restriction: For Research Use Only.
Catalog No. 50-229-2681