Learn More
Biomatik Corporation Pig Monocyte Chemotactic Protein 2 (MCP2) ELISA Kit
Supplier:  Biomatik Corporation EKU05992
Encompass_Preferred
Pig Monocyte Chemotactic Protein 2 (MCP2) ELISA Kit for detecting MCP2 in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 7.81-500pg/ml. Sensitivity: 3.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49873. Target Synonyms: CCL8; CCL-8; SCYA8; MCP-2; HC14; Chemokine C-C-Motif Ligand 8; Small Inducible Cytokine Subfamily A (Cys-Cys) Member 8; Monocyte Chemoattractant Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2683