Learn More
Biomatik Corporation Rat Myelin Oligodendrocyte Glycoprotein (MOG) ELISA Kit
Supplier:  Biomatik Corporation EKU06055
Encompass_Preferred
Rat Myelin Oligodendrocyte Glycoprotein (MOG) ELISA Kit for detecting MOG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63345. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2688