Learn More
Biomatik Corporation Human Myosin Light Chain 3, Alkali, Ventricular, Slow Skeletal (MYL3) ELISA Kit
Supplier:  Biomatik Corporation EKU06107
Encompass_Preferred
Human Myosin Light Chain 3, Alkali, Ventricular, Slow Skeletal (MYL3) ELISA Kit for detecting MYL3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08590. Gene ID: 4634. Target Synonyms: CMH8; MLC1V; VLC1; MLC1SB; Cardiac myosin light chain 1; Ventricular/slow twitch myosin alkali light chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2697