Learn More
Biomatik Corporation Mouse Meningioma Expressed Antigen 5 (MGEA5) ELISA Kit
Supplier:  Biomatik Corporation EKU06130
Encompass_Preferred
Sample Type: Serum, plasma, urine, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 3.12-200ng/mL. Sensitivity: 1.28ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9EQQ9. Target Synonyms: NAGase; MEA5; NCOAT; Hexosaminidase C; N-Acetylhexosaminidase; Hyaluronidase; Meningioma-Expressed Antigen 5; Nuclear Cytoplasmic O-GlcNAcase And Acetyltransferase. Restriction: For Research Use Only.
Catalog No. 50-229-2701