Learn More
Biomatik Corporation Human Neurofibromin 2 (NF2) ELISA Kit
Supplier:  Biomatik Corporation EKU06211
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.117ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P35240. Target Synonyms: ACN; BANF; SCH; Merlin; Bilateral Acoustic Neuroma; Schwannomin; Schwannomerlin; Moesin-Ezrin-Radixin-Like Protein. Restriction: For Research Use Only.
Catalog No. 50-229-2717