Learn More
Biomatik Corporation Rat Neutral Sphingomyelinase (NSMASE) ELISA Kit
Supplier:  Biomatik Corporation EKU06278
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.059ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9ET64. Target Synonyms: SMPD2; N-SMase; ISC1; Neutral Sphingomyelin Phosphodiesterase; Sphingomyelin Phosphodiesterase 2,Neutral Membrane; Lyso-platelet-activating factor-phospholipase C. Restriction: For Research Use Only.
Catalog No. 50-229-2725